Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004770.2 | NP_004761.2 | 2 | 1010 | ||
GenBank mRNA | AF338730.1 | - | 2 | 1010 | ||
AF450111.1 | - | 1 | 422 | |||
U69962.1 | - | 1 | 424 |