Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019741.3 | NP_062715.2 | 14 | 2948 | ||
GenBank mRNA | AK029720.1 | - | 14 | 2949 |