Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019777.3 | NP_062751.2 | 3, 4 | 429 | ||
GenBank mRNA | AB016589.1 | - | 1, 2 | 117 | ||
AK088580.1 | - | 3, 4 | 431 | |||
AK149911.1 | - | 3, 4 | 418 | |||
AK153046.1 | - | 3, 4 | 430 | |||
BC037446.1 | - | 2, 3 | 173 | 3980145 | ||
CT010206.1 | - | 1, 2 | 83 | |||
CT010313.1 | - | 1, 2 | 83 |