Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019800.4 | NP_062774.2 | 3 | 756 | ||
GenBank mRNA | AB030038.1 | - | 3 | 749 | ||
AF216223.1 | - | 3 | 233 | |||
AK002783.1 | - | 3 | 670 | |||
AK077729.1 | - | 3 | 744 | |||
AK160271.1 | - | 3 | 675 | |||
AK165356.1 | - | 3 | 757 | |||
AK171757.1 | - | 3 | 671 | |||
AK191871.1 | - | 2 | 56 | |||
BC047276.1 | - | 3 | 496 | 6487551 |