Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167799.1 | NP_001161271.1 | 3, 4 | 475 | ||
NM_019829.4 | NP_062803.4 | 2, 3 | 774 | |||
GenBank mRNA | AF232709.1 | - | 2, 3 | 142 | ||
AK002722.1 | - | 3, 4 | 457 | |||
AK135445.1 | - | 1 | 1283 | |||
AK166087.1 | - | 2, 3 | 777 | |||
AK170893.1 | - | 1 | 1578 | |||
BC004849.1 | - | 3, 4 | 408 | 2655845 | ||
BC021883.1 | - | 2, 3 | 757 | 3487476 | ||
CN539678.1 | - | 3, 4 | 439 |