Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019786.4 | NP_062760.3 | 3, 4 | 392 | ||
GenBank mRNA | AF145705.1 | - | 2, 3 | 244 | ||
AF191839.1 | - | 2, 3 | 271 | |||
AK004649.2 | - | 3, 4 | 346 | |||
AK083214.1 | - | 3, 4 | 339 | |||
AK168193.1 | - | 3, 4 | 378 | |||
BC129910.1 | - | 2, 3 | 304 | 40111752 | ||
BC129911.1 | - | 2, 3 | 304 | 40111754 | ||
BU708441.1 | - | 3, 4 | 385 |