Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 24 | 4660 | ||
NM_019831.3 | NP_062805.1 | 25 | 4796 | |||
GenBank mRNA | AF156605.1 | - | 25 | 5422 | ||
AK122268.1 | - | 23 | 5312 | |||
BC050131.1 | - | 24 | 4543 |