Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 6 | 1191 | ||
GenBank mRNA | AF021833.1 | - | 3 | 545 | ||
AK045234.1 | - | 1 | 1331 | |||
AK049399.1 | - | 6 | 1160 | |||
AK083255.1 | - | 6 | 1160 | |||
BC038483.1 | - | 3 | 249 | |||
BC054105.1 | - | 3 | 494 | 30022728 |