Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papolb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019943.2 | NP_064327.1 | 1 | 420 | ||
GenBank mRNA | AB030729.1 | - | 1 | 420 | ||
AB221598.1 | - | 1 | 230 | |||
AF039957.1 | - | 1 | 369 | |||
BC129896.1 | - | 1 | 398 | 40111521 | ||
BC129897.1 | - | 1 | 398 | 40111523 |