Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cript | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019936.3 | NP_064320.1 | 3, 4 | 244 | ||
GenBank mRNA | AF151637.1 | - | 3, 4 | 209 | ||
AK004828.1 | - | 3, 4 | 237 | |||
AK008523.1 | - | 3, 4 | 229 | |||
AK029409.1 | - | 3, 4 | 232 | |||
AK213321.1 | - | 1, 2 | 76 | |||
BC002258.1 | - | 2, 3 | 126 | |||
BC023013.1 | - | 3, 4 | 218 | 5369938 | ||
BY123761.1 | - | 3, 4 | 245 |