Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034837.2 | NP_001030009.1 | 3 | 740 | ||
NM_001034838.2 | NP_001030010.1 | 2 | 587 | |||
NM_001278339.1 | NP_001265268.1 | 2 | 707 | |||
NM_001278340.1 | NP_001265269.1 | 3 | 869 | |||
NM_014592.3 | NP_055407.1 | 2 | 707 | |||
XM_017009407.1 | XP_016864896.1 | 3 | 711 | |||
GenBank mRNA | AF199597.1 | - | 2 | 170 | ||
AI884645.1 | - | 2 | 473 | |||
AK074390.1 | - | 2 | 303 | |||
AK301775.1 | - | 2 | 362 | |||
AY170821.1 | - | 2 | 383 | |||
AY780424.1 | - | 3 | 545 | |||
BC035032.1 | - | 2 | 712 | |||
BC050375.1 | - | 2 | 474 | 5752207 | ||
BF514334.1 | - | 2 | 506 | |||
DQ148476.1 | - | 2 | 197 | |||
DQ148477.1 | - | 2 | 170 | |||
DQ148478.1 | - | 3 | 203 | |||
DQ148479.1 | - | 2 | 170 |