Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039509.1 | NP_001034598.1 | 5 | 525 | ||
NM_019999.2 | NP_064383.1 | 4 | 434 | |||
GenBank mRNA | AB041609.1 | - | 4 | 435 | ||
AK041238.1 | - | 4 | 368 | |||
AK173125.1 | - | 5 | 499 | |||
BC058945.1 | - | 6 | 621 | 6812127 |