Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020046.3 | NP_064430.1 | 2, 3 | 528 | ||
GenBank mRNA | AF029667.2 | - | 2, 3 | 233 | ||
AK013102.1 | - | 2, 3 | 480 | |||
AK030798.1 | - | 18 | ||||
AK088863.1 | - | 2, 3 | 250 | |||
AK149350.1 | - | 2, 3 | 276 | |||
BC019542.1 | - | 2, 3 | 238 | 4039461 | ||
BC027829.1 | - | 2, 3 | 489 | 5253723 | ||
BC045206.1 | - | 2, 3 | 478 | 5253764 |