Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128588.3 | NP_001122060.3 | 5, 6 | 734 | ||
NM_001146036.2 | NP_001139508.2 | 5, 6 | 663 | |||
NM_001146037.1 | NP_001139509.1 | 3, 4 | 831 | |||
NM_001308278.1 | NP_001295207.1 | 3, 4 | 408 | |||
NM_015865.6 | NP_056949.4 | 4, 5 | 576 | |||
XM_005258329.1 | XP_005258386.1 | 5, 6 | 710 | |||
XM_006722526.3 | XP_006722589.1 | 5, 6 | 711 | |||
XM_011526142.1 | XP_011524444.1 | 4, 5 | 1491 | |||
XM_011526144.1 | XP_011524446.1 | 5, 6 | 710 | |||
GenBank mRNA | AK091064.1 | - | 3, 4 | 1059 | ||
AK123681.1 | - | 2, 3 | 1635 | |||
AK289608.1 | - | 5, 6 | 677 | |||
AK294129.1 | - | 3, 4 | 347 | |||
BC050539.1 | - | 4, 5 | 502 | 5162971 | ||
GQ502682.1 | - | 2, 3 | 365 | |||
HQ709264.1 | - | 2, 3 | 344 | |||
KC677694.1 | - | 2, 3 | 344 | |||
L36121.1 | - | 2, 3 | 362 | |||
U35735.1 | - | 4, 5 | 512 | |||
Y19039.1 | - | 2, 3 | 365 |