Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114086.1 | NP_001107558.1 | 6 | 1440 | ||
NM_016929.4 | NP_058625.2 | 6 | 1204 | |||
NR_045672.1 | 3 | 782 | ||||
NR_045673.1 | 5 | 856 | ||||
XM_017010953.1 | XP_016866442.1 | 7 | 1685 | |||
GenBank mRNA | AF176920.1 | - | 6 | 1003 | ||
AF216941.1 | - | 6 | 1154 | |||
AK075163.1 | - | 6 | 1055 | |||
AK097048.1 | - | 6 | 1440 | |||
AK222544.1 | - | 6 | 1172 | |||
AK223130.1 | - | 6 | 1175 | |||
BC035968.1 | - | 6 | 1172 | 4611102 | ||
DQ679794.1 | - | 6 | 1172 |