Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006039.4 | NP_006030.2 | 2, 3 | 932 | ||
XM_011525543.2 | XP_011523845.1 | 2, 3 | 663 | |||
GenBank mRNA | AB014609.2 | - | 2, 3 | 932 | ||
AF107292.1 | - | 2, 3 | 573 | |||
AF134838.1 | - | 2, 3 | 654 | |||
BC146647.1 | - | 2, 3 | 651 | 40147137 | ||
BC150212.1 | - | 2, 3 | 636 | 40148946 | ||
BC153884.1 | - | 2, 3 | 932 | 8860500 |