Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128080.2 | NP_001121552.1 | 8 | 769 | ||
NM_001128081.1 | NP_001121553.1 | 6 | 800 | |||
NM_001128082.1 | NP_001121554.1 | 5 | 698 | |||
NM_020286.3 | NP_064682.1 | 7 | 884 | |||
GenBank mRNA | AF291425.1 | - | 7 | 852 | ||
AJ279791.1 | - | 7 | 844 | |||
AJ279792.1 | - | 6 | 760 | |||
AJ279794.1 | - | 5 | 658 | |||
AK013248.1 | - | 6 | 575 | |||
AK154694.1 | - | 7 | 905 | |||
AK171199.1 | - | 7 | 989 | |||
AY039000.1 | - | 8 | 777 | |||
BC051204.1 | - | 5 | 688 | |||
BC055858.1 | - | 6 | 774 | 4913671 |