Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159502.1 | NP_001152974.1 | 3, 4 | 1511 | ||
NM_020494.3 | NP_065240.2 | 3, 4 | 1345 | |||
GenBank mRNA | AF214732.1 | - | 3, 4 | 1325 | ||
AK134339.1 | - | 3, 4 | 1323 | |||
AK149810.1 | - | 3, 4 | 1297 | |||
AK150778.1 | - | 3, 4 | 1297 | |||
AK152551.1 | - | 3, 4 | 1297 | |||
AK152928.1 | - | 3, 4 | 1297 | |||
AK166360.1 | - | 3, 4 | 1306 | |||
AK172557.1 | - | 3, 4 | 1298 | |||
BC043721.1 | - | 3, 4 | 1313 | |||
BC055048.1 | - | 3, 4 | 1275 | 6484820 | ||
BC055317.1 | - | 3, 4 | 1511 | 5696084 | ||
U46690.1 | - | 1, 2 | 59 |