Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020581.2 | NP_065606.2 | 2 | 533 | ||
GenBank mRNA | AB054540.1 | - | 2 | 419 | ||
AF123261.1 | - | 2 | 464 | |||
AF169313.1 | - | 2 | 396 | |||
AF278699.1 | - | 2 | 468 | |||
AK009161.1 | - | 2 | 239 | |||
AK014564.1 | - | 534 | ||||
AK032491.1 | - | 2 | 502 | |||
AK132761.1 | - | 2 | 502 | |||
AK134419.1 | - | 2 | 502 | |||
BC006611.1 | - | 2 | 485 | 3966729 | ||
BC021343.1 | - | 2 | 499 | 4194173 | ||
BC025797.1 | - | 2 | 469 | 5137159 | ||
KJ650202.1 | - | 2 | 350 |