Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173483.1 | NP_001166954.1 | 4, 5 | 626 | ||
NM_020599.2 | NP_065624.1 | 4, 5 | 708 | |||
GenBank mRNA | AK014227.1 | - | 4, 5 | 399 | ||
AK140622.1 | - | 4, 5 | 627 | |||
BC016268.1 | - | 3, 4 | 301 | 4481973 |