Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020587.2 | NP_065612.2 | 6 | 1506 | ||
GenBank mRNA | AB041587.1 | - | 3 | 944 | ||
AK075769.1 | - | 6 | 1471 | |||
AK077648.1 | - | 6 | 1492 | |||
AK156989.1 | - | 6 | 1441 | |||
BC019437.1 | - | 6 | 1460 | 4018178 | ||
BC026944.1 | - | 6 | 2366 | 4016087 | ||
BC040812.1 | - | 6 | 2366 |