Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021378.1 | NP_067353.1 | 3 | 292 | ||
GenBank mRNA | AF195779.1 | - | 3 | 292 | ||
BC130234.1 | - | 4 | 371 | 40142004 |