Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322080.1 | NP_001309009.1 | 4, 5 | 629 | ||
NM_001322081.1 | NP_001309010.1 | 4, 5 | 530 | |||
NM_032718.4 | NP_116107.3 | 4, 5 | 513 | |||
XM_011512005.2 | XP_011510307.1 | 4, 5 | 636 | |||
GenBank mRNA | AK075376.1 | - | 4, 5 | 479 | ||
AK290589.1 | - | 4, 5 | 467 | |||
AK296768.1 | - | 4, 5 | 629 | |||
BC006242.2 | - | 4, 5 | 482 | 3952602 |