Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021387.3 | NP_067362.1 | 2, 3 | 365 | ||
GenBank mRNA | AB030198.1 | - | 2, 3 | 366 | ||
AK013845.1 | - | 2, 3 | 353 |