Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004468.2 | NP_001004468.1 | 17 | 8107 | ||
NM_021314.4 | NP_067289.2 | 13 | 2942 | |||
NM_206856.3 | NP_996738.2 | 16 | 2908 | |||
GenBank mRNA | AB041546.1 | - | 13 | 2943 | ||
AK050985.1 | - | 11 | 2570 | |||
AK145721.1 | - | 11 | 2571 | |||
AK165194.1 | - | 11 | 2756 | |||
AK165603.1 | - | 10 | 2466 | |||
AY177410.1 | - | 15 | 2920 | |||
BC004057.1 | - | 8 | 1982 |