Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012471.2 | NP_036603.1 | 2 | 1086 | ||
XM_017029774.1 | XP_016885263.1 | 3 | 1376 | |||
GenBank mRNA | AB209258.1 | - | 2 | 500 | ||
AF054568.1 | - | 2 | 1085 | |||
BC137271.1 | - | 2 | 926 | 9021268 | ||
BC137274.1 | - | 2 | 926 | 9021271 |