Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqrdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162503.1 | NP_001155975.1 | 2, 3 | 342 | ||
NM_021507.5 | NP_067482.4 | 2, 3 | 661 | |||
GenBank mRNA | AF174535.1 | - | 2, 3 | 661 | ||
AK008290.1 | - | 2, 3 | 308 | |||
AK016166.1 | - | 2, 3 | 294 | |||
AK075598.1 | - | 2, 3 | 343 | |||
AK149948.1 | - | 2, 3 | 313 | |||
BC011153.1 | - | 1, 2 | 280 | 4196280 | ||
BC028247.1 | - | 2, 3 | 330 |