Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamdec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021475.2 | NP_067450.1 | 11 | 1114 | ||
GenBank mRNA | AJ242912.1 | - | 11 | 1114 | ||
AK008929.1 | - | 2 | 125 | |||
AK156586.1 | - | 11 | 1129 | |||
BC026542.1 | - | 11 | 1138 | |||
BC046324.1 | - | 11 | 1105 | 6394673 |