Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163487.1 | NP_001156959.1 | 6 | 604 | ||
NM_001163488.1 | NP_001156960.1 | 6 | 540 | |||
NM_021514.4 | NP_067489.3 | 5 | 557 | |||
GenBank mRNA | AF249894.1 | - | 5 | 497 | ||
AK002711.2 | - | 6 | 507 | |||
AK049773.1 | - | 6 | 512 | |||
BC005526.1 | - | 5 | 521 | 3495173 | ||
BC115774.1 | - | 3 | 312 | |||
CX218942.1 | - | 6 | 596 | |||
D21865.1 | - | 5 | 455 |