Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021555.2 | NP_067530.2 | 3, 4 | 867 | ||
GenBank mRNA | AB041552.1 | - | 3, 4 | 867 | ||
AK085815.1 | - | 3, 4 | 861 | |||
AK178524.1 | - | 1, 2 | 146 | |||
BC061251.1 | - | 3, 4 | 831 | 30249656 |