Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160130.1 | NP_001153602.1 | 3 | 932 | ||
NM_001160132.1 | NP_001153604.1 | 3 | 932 | |||
NM_001160133.1 | NP_001153605.1 | 3 | 932 | |||
NM_001160134.1 | NP_001153606.1 | 3 | 932 | |||
NM_019842.3 | NP_062816.2 | 3 | 932 | |||
XM_017011058.1 | XP_016866547.1 | 3 | 932 | |||
GenBank mRNA | AF202977.1 | - | 3 | 480 | ||
AF249278.1 | - | 3 | 694 | |||
AF263835.1 | - | 3 | 372 | |||
AK299550.1 | - | 3 | 532 | |||
BC050689.1 | - | 3 | 669 | |||
BC117359.1 | - | 3 | 603 | 40125910 | ||
BC143554.1 | - | 3 | 603 | 9052065 |