Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159631.1 | NP_001153103.1 | 3 | 315 | ||
NM_021606.3 | NP_067619.1 | 3 | 308 | |||
GenBank mRNA | AF218847.1 | - | 3 | 310 | ||
AK004925.1 | - | 3 | 285 | |||
AK011519.1 | - | 3 | 294 | |||
AK018168.1 | - | 3 | 298 | |||
AK029266.1 | - | 3 | 297 | |||
AK075717.1 | - | 3 | 282 | |||
AK075836.1 | - | 3 | 294 | |||
AK086700.1 | - | 313 | ||||
AK089919.1 | - | 3 | 269 | |||
AK170757.1 | - | 3 | 315 | |||
BC019524.1 | - | 3 | 377 | 4209097 | ||
BY205493.1 | - | 3 | 315 |