Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021790.2 | NP_068562.1 | 7 | 764 | ||
NM_181061.5 | NP_851406.1 | 7 | 887 | |||
GenBank mRNA | AB043687.1 | - | 7 | 764 | ||
AK082939.1 | - | 7 | 544 | |||
AK135773.1 | - | 280 | ||||
AK160776.1 | - | 7 | 535 | |||
AK165268.1 | - | 2 | 2971 | |||
BC028655.1 | - | 7 | 900 |