Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199105.1 | NP_001186034.1 | 3 | 541 | ||
NM_021897.3 | NP_068697.1 | 3 | 541 | |||
GenBank mRNA | AJ131776.1 | - | 3 | 542 | ||
AK054214.1 | - | 3 | 538 | |||
AK142321.1 | - | 3 | 537 | |||
AY034611.1 | - | 3 | 542 | |||
AY034612.1 | - | 3 | 542 | |||
BC132124.1 | - | 2 | 443 | 40130401 | ||
BC138711.1 | - | 2 | 443 | 8861733 | ||
BC145396.1 | - | 2 | 442 |