Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022029.2 | NP_071312.1 | 2 | 330 | ||
GenBank mRNA | AK002933.1 | - | 2 | 290 | ||
AK158630.1 | - | 2 | 317 | |||
BC061102.1 | - | 2 | 294 | 6590375 | ||
BC138511.1 | - | 2 | 207 | 8861531 | ||
BC138513.1 | - | 2 | 207 | 8861533 | ||
BU604757.1 | - | 2 | 333 |