Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013437.1 | NP_001013455.1 | 4 | 463 | ||
NM_031216.3 | NP_112493.2 | 4 | 463 | |||
XM_005258152.4 | XP_005258209.1 | 3 | 306 | |||
XM_011525742.2 | XP_011524044.1 | 4 | 459 | |||
XM_011525743.2 | XP_011524045.1 | 3 | 306 | |||
XM_011525744.1 | XP_011524046.1 | 4 | 459 | |||
XM_011525745.2 | XP_011524047.1 | 3 | 227 | |||
XM_011525746.2 | XP_011524048.1 | 4 | 569 | |||
XM_017026025.1 | XP_016881514.1 | 3 | 312 | |||
XM_017026026.1 | XP_016881515.1 | 3 | 383 | |||
XM_017026027.1 | XP_016881516.1 | 4 | 633 | |||
GenBank mRNA | AF136976.1 | - | 4 | 431 | ||
AF255625.1 | - | 4 | 367 | |||
AF431970.1 | - | 4 | 367 | |||
AF514996.1 | - | 4 | 327 | |||
AK056940.1 | - | 4 | 426 | |||
AK291226.1 | - | 4 | 426 | |||
AK314403.1 | - | 4 | 437 | |||
BC012430.1 | - | 4 | 417 | 3865994 | ||
CB990796.1 | - | 490 |