Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020044.3 | NP_064428.1 | 2, 3 | 268 | ||
NM_022964.4 | NP_075253.2 | 2, 3 | 268 | |||
GenBank mRNA | AF257136.1 | - | 1, 2 | 212 | ||
AK138953.1 | - | 2, 3 | 227 | |||
AK143533.1 | - | 2, 3 | 269 | |||
AK144261.1 | - | 2, 3 | 207 | |||
AK162321.1 | - | 2, 3 | 227 | |||
AY190024.1 | - | 2, 3 | 233 | |||
BC005804.1 | - | 2, 3 | 197 | 3154073 | ||
BY228290.1 | - | 2, 3 | 267 |