Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022315.2 | NP_071710.2 | 3 | 527 | ||
GenBank mRNA | AJ249901.1 | - | 3 | 498 | ||
AK030958.1 | - | 3 | 496 | |||
AK159749.1 | - | 3 | 494 | |||
AK160002.1 | - | 3 | 494 | |||
AK172312.1 | - | 3 | 492 | |||
BC019527.1 | - | 3 | 474 | 4191849 |