Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 9 | 986 | ||
NM_001199114.1 | NP_001186043.1 | 8 | 825 | |||
NM_001199115.1 | NP_001186044.1 | 8 | 931 | |||
NM_001199116.1 | NP_001186045.1 | 8 | 891 | |||
NM_022880.3 | NP_075018.1 | 7 | 932 | |||
GenBank mRNA | AF131212.1 | - | 7 | 933 | ||
AF305501.1 | - | 8 | 896 | |||
AK004756.1 | - | 8 | 877 | |||
AK050089.1 | - | 8 | 916 | |||
AK146265.1 | - | 8 | 826 | |||
AK149483.1 | - | 8 | 847 | |||
AK170463.1 | - | 8 | 863 | |||
BC004828.1 | - | 9 | 949 | 2631728 | ||
BC006812.1 | - | 7 | 863 | 3155188 | ||
EU180577.1 | - | 7 | 801 |