Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130770.2 | NP_570126.2 | 6 | 630 | ||
GenBank mRNA | AF459285.1 | - | 6 | 630 | ||
BC131799.1 | - | 6 | 621 | 40069671 |