Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301137.1 | NP_001288066.1 | 6 | 2315 | ||
NM_022428.3 | NP_071873.2 | 6 | 2318 | |||
GenBank mRNA | AJ271055.1 | - | 6 | 1579 | ||
AK035140.1 | - | 6 | 2321 | |||
AK053729.1 | - | 6 | 2206 | |||
BC036969.1 | - | 3 | 909 | |||
BU054800.1 | - | 2 | 586 |