Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042659.1 | NP_001036124.1 | 2 | 1835 | ||
NM_022721.3 | NP_073558.2 | 2 | 1942 | |||
GenBank mRNA | AB052910.1 | - | 1 | 1068 | ||
AF272146.1 | - | 1 | 1059 | |||
AK030111.1 | - | 2 | 1944 | |||
AK039402.1 | - | 2 | 1835 | |||
BC117723.2 | - | 2 | 1368 | 40088671 |
Set Membership: |
G-protein coupled receptor |