Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023049.1 | NP_075536.1 | 2 | 581 | ||
GenBank mRNA | AB221595.1 | - | 1 | 191 | ||
BC031161.1 | - | 2 | 581 | 4981777 |