Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023053.3 | NP_075540.1 | 4 | 538 | ||
GenBank mRNA | AF292033.1 | - | 477 | |||
AJ297390.1 | - | 4 | 488 | |||
AK033504.1 | - | 4 | 497 | |||
AK049646.1 | - | 4 | 532 | |||
AK168547.1 | - | 4 | 488 | |||
AK168633.1 | - | 4 | 497 | |||
BC004850.1 | - | 4 | 493 | 2810960 |