Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023119.2 | NP_075608.2 | 3, 4 | 341 | ||
GenBank mRNA | AK002322.1 | - | 3, 4 | 300 | ||
AK002336.1 | - | 3, 4 | 299 | |||
AK088788.1 | - | 3, 4 | 285 | |||
AK147987.1 | - | 3, 4 | 299 | |||
AK150161.1 | - | 3, 4 | 299 | |||
AK150192.1 | - | 3, 4 | 300 | |||
AK153321.1 | - | 3, 4 | 298 | |||
AK180361.1 | - | 1, 2 | 23 | |||
AK191162.1 | - | 23 | ||||
AK193395.1 | - | 23 | ||||
AK199229.1 | - | 1, 2 | 23 | |||
AK202164.1 | - | 1, 2 | 23 | |||
AK202428.1 | - | 1, 2 | 23 | |||
AK203149.1 | - | 1, 2 | 23 | |||
AK204087.1 | - | 1, 2 | 23 | |||
AK204457.1 | - | 1, 2 | 23 | |||
AK204949.1 | - | 1, 2 | 23 | |||
AK214077.1 | - | 2, 3 | 157 | |||
BC003891.1 | - | 3, 4 | 291 | 3592683 | ||
BC003934.1 | - | 3, 4 | 313 | |||
BC010685.1 | - | 3, 4 | 279 | 3599051 | ||
BC024644.1 | - | 3, 4 | 271 | 3583123 | ||
BC039179.1 | - | 3, 4 | 261 | 4501041 | ||
BC083334.1 | - | 3, 4 | 273 | 6414729 | ||
BC085098.1 | - | 2, 3 | 202 | 5376359 | ||
BU526382.1 | - | 3, 4 | 344 | |||
X52379.1 | - | 3, 4 | 283 |
Gene Symbol: Eno1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025388.1 | NP_001020559.1 | 2 | 323 | ||
GenBank mRNA | BC089539.1 | - | 2 | 323 | 6707317 |