Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081682.2 | NP_001075151.1 | 3 | 361 | ||
NM_001285491.1 | NP_001272420.1 | 3 | 361 | |||
NM_001285493.1 | NP_001272422.1 | 3 | 303 | |||
NM_001285494.1 | NP_001272423.1 | 3 | 303 | |||
NM_023120.5 | NP_075609.2 | 3 | 303 | |||
GenBank mRNA | AF301595.1 | - | 3 | 254 | ||
AK010489.1 | - | 3 | 261 | |||
AK016952.1 | - | 3 | 291 | |||
AK087954.1 | - | 3 | 360 | |||
AK158210.1 | - | 3 | 290 | |||
AK166195.1 | - | 3 | 287 | |||
AK171253.1 | - | 3 | 283 | |||
AK173227.1 | - | 3 | 286 | |||
BC024635.1 | - | 3 | 272 | 4193427 | ||
BC037676.1 | - | 2 | 189 | 3493788 | ||
BY328275.1 | - | 3 | 309 |
Set Membership: |
G-protein coupled receptor |