Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258459.1 | NP_001245388.1 | 6 | 656 | ||
NM_001258460.1 | NP_001245389.1 | 5 | 533 | |||
NM_006213.4 | NP_006204.1 | 5 | 560 | |||
NR_047689.1 | 4 | 505 | ||||
XM_005271772.4 | XP_005271829.1 | 3 | 777 | |||
XM_017012324.1 | XP_016867813.1 | 5 | 560 | |||
XM_017012325.1 | XP_016867814.1 | 2 | 221 | |||
XM_017012326.1 | XP_016867815.1 | 3 | 780 | |||
XM_017012327.1 | XP_016867816.1 | 2 | 189 | |||
GenBank mRNA | AK293180.1 | - | 6 | 611 | ||
AK299429.1 | - | 4 | 461 | |||
AK300928.1 | - | 5 | 485 | |||
AK312851.1 | - | 5 | 560 | |||
BC051327.1 | - | 5 | 498 | 6277230 | ||
BC069679.1 | - | 5 | 476 | 7262511 | ||
BC069738.1 | - | 5 | 477 | 7262523 | ||
BC069754.1 | - | 5 | 477 | 7262535 | ||
BC074753.2 | - | 4 | 404 | 30915212 | ||
DB171823.1 | - | 5 | 560 | |||
X80590.1 | - | 5 | 484 |