Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204959.1 | NP_001191888.1 | 4 | 388 | ||
NM_022984.4 | NP_075360.1 | 4 | 388 | |||
GenBank mRNA | AF290870.1 | - | 4 | 381 | ||
AF323080.1 | - | 4 | 387 | |||
AK131613.1 | - | 4 | 389 | |||
AK131869.1 | - | 4 | 385 | |||
BC051196.1 | - | 4 | 374 | 1448672 |