Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023049.1 | NP_075536.1 | 4 | 754 | ||
GenBank mRNA | AB221595.1 | - | 3 | 364 | ||
AF155353.1 | - | 2 | 221 | |||
AK083400.1 | - | 2 | 280 | |||
BC031161.1 | - | 4 | 754 | 4981777 |