Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023060.3 | NP_075547.1 | 5 | 1394 | ||
GenBank mRNA | AF268871.1 | - | 5 | 1358 | ||
AF283518.1 | - | 5 | 1349 | |||
AK028424.1 | - | 5 | 1376 | |||
AK087501.1 | - | 5 | 1382 | |||
AK147924.1 | - | 5 | 1363 | |||
AK171885.1 | - | 5 | 1374 | |||
BC024915.1 | - | 5 | 1368 | 3158026 |